Mannose triflate
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose, TATM
- CAS Number:92051-23-5
- Formula:C₁₅H₁₉F₃O₁₂S
- MDL Number:MFCD00012353
- Molecular Weight:480.36 g/mol
- Melting Pt:118…122 °C
- Storage Temperature:Freezer
5 Options Available Below
Product Details & Documents
Synonyms: (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose, TATM
- CAS Number:92051-23-5
- Formula:C₁₅H₁₉F₃O₁₂S
- MDL Number:MFCD00012353
- Molecular Weight:480.36 g/mol
- Melting Pt:118…122 °C
- Storage Temperature:Freezer
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



