Order Entry
Export
ContactUsLinkComponent
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluoro-14-iodotetradecane 90%
Irritant
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluoro-14-iodotetradecane 90%
Catalog # APOSPC9868-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluoro-14-iodotetradecane 90%
Catalog # APOSPC9868-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :774.05 g/mol
Melting Pt:110…113 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:30046-31-2
Formula:C₁₄H₄F₂₅I