Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,3-Difluoropyrrolidine hydrochloride
3,3-Difluoropyrrolidine hydrochloride
3,3-Difluoropyrrolidine hydrochloride
Irritant
3,3-Difluoropyrrolidine hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,3-Difluoropyrrolidine hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
163457-23-6
:
Formula:
C₄H₇F₂N
MDL Number:
MFCD03788948
:
143.56 g/mol
Melting Pt:
128…134 °C
:
GHS07
Storage Temperature:
Ambient