0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3-Chloro-2,6-difluoroanisole
3-Chloro-2,6-difluoroanisole 98%
3-Chloro-2,6-difluoroanisole 98%
APOSPC9732-1G
:
Apollo Scientific
:
3-Chloro-2,6-difluoroanisole 98%
APOSPC9732-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
178.57 g/mol
:
GHS07
:
Ambient
:
261762-38-3
:
C₇H₅ClF₂O
:
MFCD01631331