

3,5-Bis(trifluoromethyl)anisole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:349-60-0
:
Formula:C₉H₆F₆O
MDL Number:MFCD00728736
:244.14 g/mol
Boiling Pt:158…160 °C
Flash Pt:55 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07