6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338783-19-0
:
Formula:C₁₄H₁₀F₃NO₃
MDL Number:MFCD00139896
:297.24 g/mol
Melting Pt:204 °C
Storage Temperature:Ambient