
2,3-Difluoro-6-nitroaniline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:211693-73-1
:
Formula:C₆H₄F₂N₂O₂
MDL Number:MFCD00233313
:174.11 g/mol
Melting Pt:114…118 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator