Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluoronicotinic acid
2,6-Difluoronicotinic acid 96%
2,6-Difluoronicotinic acid 96%
Irritant
2,6-Difluoronicotinic acid 96%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,6-Difluoronicotinic acid 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
171178-50-0
:
Formula:
C₆H₃F₂NO₂
MDL Number:
MFCD08064047
:
159.0900 g/mol
Melting Pt:
165…167 °C
:
GHS07
Storage Temperature:
Ambient