Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3,4-Difluorophenoxy)acetic acid
(3,4-Difluorophenoxy)acetic acid 98%
(3,4-Difluorophenoxy)acetic acid 98%
(3,4-Difluorophenoxy)acetic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
(3,4-Difluorophenoxy)acetic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
370-58-1
:
Formula:
C₈H₆F₂O₃
MDL Number:
MFCD06823827
:
188.13 g/mol
Storage Temperature:
Ambient