Order Entry
Export
ContactUsLinkComponent
4-Chloro-2,5-difluoroanisole 95%
4-Chloro-2,5-difluoroanisole 95%
Catalog # APOSPC910099-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-2,5-difluoroanisole 95%
Catalog # APOSPC910099-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:1261846-22-3
Formula:C₇H₅ClF₂O