Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-3,5-dichlorofluorobenzene
4-Bromo-3,5-dichlorofluorobenzene 95%
4-Bromo-3,5-dichlorofluorobenzene 95%
Irritant
4-Bromo-3,5-dichlorofluorobenzene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Bromo-3,5-dichlorofluorobenzene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
263333-82-0
:
Formula:
C₆H₂BrCl₂F
MDL Number:
MFCD11845890
:
243.9 g/mol
:
GHS07
Storage Temperature:
Ambient