Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Difluoro-6-methylpyridine
2,3-Difluoro-6-methylpyridine 98%
2,3-Difluoro-6-methylpyridine 98%
2,3-Difluoro-6-methylpyridine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Difluoro-6-methylpyridine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1227579-04-5
:
Formula:
C₆H₅F₂N
MDL Number:
MFCD16610828
:
129.11 g/mol
Storage Temperature:
Ambient