Order Entry
Export
ContactUsLinkComponent
2,6-Dimethyl-4-fluorobenzonitrile 95%
2,6-Dimethyl-4-fluorobenzonitrile 95%
Catalog # APOSPC905491-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dimethyl-4-fluorobenzonitrile 95%
Catalog # APOSPC905491-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :149.1649 g/mol
Storage Temperature:Ambient
CAS Number:14659-61-1
Formula:C₉H₈FN
MDL Number:MFCD04972839