Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-4,4-difluorocyclohexanecarboxylate
Methyl-4,4-difluorocyclohexanecarboxylate 97%
Methyl-4,4-difluorocyclohexanecarboxylate 97%
Irritant
Methyl-4,4-difluorocyclohexanecarboxylate 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-4,4-difluorocyclohexanecarboxylate 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
121629-14-9
:
Formula:
C₈H₁₂F₂O₂
MDL Number:
MFCD11040454
:
178.18 g/mol
:
GHS07
Storage Temperature:
Refrigerator