0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2-Chloro-3,4-difluorobenzaldehyde
2-Chloro-3,4-difluorobenzaldehyde 98%
2-Chloro-3,4-difluorobenzaldehyde 98%
APOSPC903986-250MG
:
Apollo Scientific
:
2-Chloro-3,4-difluorobenzaldehyde 98%
APOSPC903986-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
176.5479 g/mol
:
GHS07
:
Refrigerator
:
1261869-07-1
:
C₇H₃ClF₂O
:
MFCD18415551