Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluoro-1,3-benzodioxole
4-Fluoro-1,3-benzodioxole 98%
4-Fluoro-1,3-benzodioxole 98%
4-Fluoro-1,3-benzodioxole 98%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Fluoro-1,3-benzodioxole 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
943830-74-8
:
Formula:
C₇H₅FO₂
MDL Number:
MFCD12026062
Storage Temperature:
Ambient