2-Bromo-3,4-difluoroanisole 95%
Catalog # APOSPC902904-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:223.148 g/mol
Storage Temperature:Ambient
CAS Number:935285-66-8
Formula:C₇H₅BrF₂O
MDL Number:MFCD09261345