Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Difluoro-2-methylbenzonitrile
4,5-Difluoro-2-methylbenzonitrile 95%
4,5-Difluoro-2-methylbenzonitrile 95%
4,5-Difluoro-2-methylbenzonitrile 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4,5-Difluoro-2-methylbenzonitrile 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1003708-82-4
:
Formula:
C₈H₅F₂N
MDL Number:
MFCD09996935
:
153.1300 g/mol
Storage Temperature:
Ambient