Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole 97%
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole 97%
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole 97%
Supplier:
Apollo Scientific
Certificates
Expand image
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
175136-60-4
:
Formula:
C₁₂H₇F₆N
MDL Number:
MFCD00053048
:
279.19 g/mol
Melting Pt:
43 °C
Storage Temperature:
Ambient