Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-4-fluorobenzo[d][1,3]dioxole
5-Bromo-4-fluorobenzo[d][1,3]dioxole 97%
5-Bromo-4-fluorobenzo[d][1,3]dioxole 97%
Irritant
5-Bromo-4-fluorobenzo[d][1,3]dioxole 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Bromo-4-fluorobenzo[d][1,3]dioxole 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1226808-75-8
:
Formula:
C₇H₄BrFO₂
MDL Number:
MFCD16295053
:
219.009 g/mol
:
GHS07
Storage Temperature:
Ambient