Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2,8-bis(trifluoromethyl)quinoline
4-Chloro-2,8-bis(trifluoromethyl)quinoline 97%
4-Chloro-2,8-bis(trifluoromethyl)quinoline 97%
:
Apollo Scientific
4-Chloro-2,8-bis(trifluoromethyl)quinoline 97%
:
Apollo Scientific
:
83012-13-9
:
:
C₁₁H₄ClF₆N
:
MFCD00075104
:
299.6 g/mol
:
46…48 °C
:
113 °C
:
GHS07
:
Ambient