
3,3'-Bis(trifluoromethyl)diphenylmethane
Catalog # APOSPC8960-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:304.24 g/mol
Boiling Pt:85…87 °C/0.5mmHg
Melting Pt:37…42 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
CAS Number:86845-35-4
Formula:C₁₅H₁₀F₆
MDL Number:MFCD00000396