
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:178488-36-3
:
Formula:C₈H₄ClF₃N₂
MDL Number:MFCD06496224
:220.58 g/mol
Melting Pt:110…112 °C
:GHS07
Storage Temperature:Ambient