0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1-n-Butoxy-2,3-difluorobenzene
1-n-Butoxy-2,3-difluorobenzene 99%
1-n-Butoxy-2,3-difluorobenzene 99%
:
Apollo Scientific
1-n-Butoxy-2,3-difluorobenzene 99%
:
Apollo Scientific
:
136239-66-2
:
:
C₁₀H₁₂F₂O
:
MFCD07368738
:
186.202 g/mol
:
GHS07
:
Ambient