0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2-(3-Fluorophenyl)-1,3-thiazole-4-carboxylic acid
2-(3-Fluorophenyl)-1,3-thiazole-4-carboxylic acid
2-(3-Fluorophenyl)-1,3-thiazole-4-carboxylic acid
2-(3-Fluorophenyl)-1,3-thiazole-4-carboxylic acid
Supplier:
Apollo Scientific
Certificates
Expand image
2-(3-Fluorophenyl)-1,3-thiazole-4-carboxylic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
886369-06-8
:
Formula:
C₁₀H₆FNO₂S
MDL Number:
MFCD07375293
:
223.23 g/mol
Melting Pt:
211…213 °C
Storage Temperature:
Ambient