Order Entry
Export
ContactUsLinkComponent
2-(4-Fluorophenyl)-4H-3,1-benzoxazin-4-one
2-(4-Fluorophenyl)-4H-3,1-benzoxazin-4-one
Catalog # APOSPC8487-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Fluorophenyl)-4H-3,1-benzoxazin-4-one
Catalog # APOSPC8487-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :241.22 g/mol
Melting Pt:180…181 °C
Storage Temperature:Ambient
CAS Number:18600-51-6
Formula:C₁₄H₈FNO₂
MDL Number:MFCD00087177