Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluorophenetole
3,4-Difluorophenetole 98%
3,4-Difluorophenetole 98%
3,4-Difluorophenetole 98%
Supplier:
Apollo Scientific
Certificates
Expand image
3,4-Difluorophenetole 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
163848-46-2
:
Formula:
C₈H₈F₂O
MDL Number:
MFCD00673005
:
158.15 g/mol
Storage Temperature:
Ambient