Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dichloro-2,4-difluorobenzene
1,3-Dichloro-2,4-difluorobenzene 97%
1,3-Dichloro-2,4-difluorobenzene 97%
:
Apollo Scientific
1,3-Dichloro-2,4-difluorobenzene 97%
:
Apollo Scientific
:
36556-37-3
:
:
C₆H₂Cl₂F₂
:
MFCD00142519
:
182.98 g/mol
:
GHS07
:
Ambient