Order Entry
Export
ContactUsLinkComponent
3-Chloro-2,5-difluoropyridine 98%
Flammable
Corrosive
Irritant
3-Chloro-2,5-difluoropyridine 98%
Catalog # APOSPC8011-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Corrosive
Irritant
3-Chloro-2,5-difluoropyridine 98%
Catalog # APOSPC8011-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :149.53 g/mol
Storage Temperature:Ambient
UN:2924
ADR:3+8,III
 :GHS02
 :GHS05
 :GHS07
CAS Number:851179-00-5
Formula:C₅H₂ClF₂N
MDL Number:MFCD09998887