2-[4-(Trifluoromethyl)piperidino]-1,3-thiazole-5-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1000339-88-7
:
Formula:C₁₀H₁₁F₃N₂OS
MDL Number:MFCD09864683
Melting Pt:42…45 °C
:264.27 g/mol
Storage Temperature:Ambient