
2,6-Difluoronicotinaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:155601-65-3
:
Formula:C₆H₃F₂NO
MDL Number:MFCD08277633
:143.093 g/mol
Boiling Pt:64…66 °C/15mmHg
Melting Pt:19…20 °C
Density:1.378 g/cm³ (25 °C)
:GHS07
Storage Temperature:Freezer