Order Entry
Export
Orders LinkContactUsLinkComponent
 

4,4-Difluorocyclohexanone

Flammable
Irritant
Supplier:   Apollo Scientific
4,4-Difluorocyclohexanone
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:22515-18-0
  • Formula:C₆H₈F₂O
  • MDL Number:MFCD08146635
  • Molecular Weight:134.12 g/mol
  • Melting Pt:34…37 °C
  • Flash Pt:56 °C
  • Storage Temperature:Ambient
  • UN:1325
  • ADR:4.1,III
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 43,50 €
    43,50 €
    1 * 1 g
       43,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSPC7922-1G
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 78,20 €
    78,20 €
    1 * 5 g
       78,20 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSPC7922-5G
  • Bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 113,00 €
    113,00 €
    1 * 10 g
       113,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSPC7922-10G