Order Entry
Export
ContactUsLinkComponent
4-Bromo-2,5-difluorophenol 98+%
Irritant
4-Bromo-2,5-difluorophenol 98+%
Catalog # APOSPC7898-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromo-2,5-difluorophenol 98+%
Catalog # APOSPC7898-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :208.99 g/mol
Melting Pt:44…53 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:486424-36-6
Formula:C₆H₃BrF₂O
MDL Number:MFCD04973751