Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromo-3-fluoro-o-xylene
6-Bromo-3-fluoro-o-xylene 97%
6-Bromo-3-fluoro-o-xylene 97%
6-Bromo-3-fluoro-o-xylene 97%
APOSPC7883-1G
:
Apollo Scientific
:
6-Bromo-3-fluoro-o-xylene 97%
APOSPC7883-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
203.06 g/mol
:
GHS07
:
Ambient
:
52548-00-2
:
C₈H₈BrF
:
MFCD03789115