Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 99+%
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 99+%
:
Apollo Scientific
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 99+%
:
Apollo Scientific
:
368-63-8
:
:
C₁₀H₈F₆O
:
MFCD01632521
:
258.16 g/mol
:
70…74 °C
:
GHS07
:
Ambient