
2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:675602-89-8
:
Formula:C₆H₃F₅N₂
MDL Number:MFCD03840200
:198.10 g/mol
Melting Pt:66…68 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07