Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Difluoro-4-methoxybenzaldehyde
3,5-Difluoro-4-methoxybenzaldehyde
3,5-Difluoro-4-methoxybenzaldehyde
Irritant
3,5-Difluoro-4-methoxybenzaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Difluoro-4-methoxybenzaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
654-11-5
:
Formula:
C₈H₆F₂O₂
MDL Number:
MFCD04115911
:
172.13 g/mol
Melting Pt:
38…41 °C
:
GHS07
Storage Temperature:
Ambient