Order Entry
Export
ContactUsLinkComponent
3,4-Difluorosalicylic acid 98%
Irritant
3,4-Difluorosalicylic acid 98%
Catalog # APOSPC7835-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,4-Difluorosalicylic acid 98%
Catalog # APOSPC7835-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.1 g/mol
Melting Pt:177…179 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:189283-51-0
Formula:C₇H₄F₂O₃
MDL Number:MFCD03428564