

α'-Chloro-α,α,α-trifluoro-o-xylene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21742-00-7
:
Formula:C₈H₆ClF₃
MDL Number:MFCD00009920
:194.58 g/mol
Boiling Pt:67…68 °C/12mmHg
Density:1.337 g/cm³ (25 °C)
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:244-560-6
UN:3265
ADR:8,III
:GHS05
:GHS07