2-Amino-4-[(3,5-difluoro)phenyl]-1,3-thiazole
Catalog # APOSPC7556-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.22 g/mol
Storage Temperature:Ambient
CAS Number:676348-23-5
Formula:C₉H₆F₂N₂S
MDL Number:MFCD03424572