
2-Amino-4-trifluoromethyl-1,3-thiazole
Catalog # APOSPC7554-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:168.14 g/mol
Melting Pt:65…70 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:349-49-5
Formula:C₄H₃F₃N₂S
MDL Number:MFCD00832759