Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Trifluoromethyl)benzhydrol
3-(Trifluoromethyl)benzhydrol 97%
3-(Trifluoromethyl)benzhydrol 97%
:
Apollo Scientific
3-(Trifluoromethyl)benzhydrol 97%
:
Apollo Scientific
:
728-80-3
:
:
C₁₄H₁₁F₃O
:
MFCD00014397
:
252.23 g/mol
:
125…127 °C/0.5mmHg
:
63…64 °C
:
GHS07
:
Ambient