Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dichloro-5-nitrobenzotrifluoride
3,4-Dichloro-5-nitrobenzotrifluoride
3,4-Dichloro-5-nitrobenzotrifluoride
:
Apollo Scientific
3,4-Dichloro-5-nitrobenzotrifluoride
:
Apollo Scientific
:
657-02-3
:
:
C₇H₂Cl₂F₃NO₂
:
MFCD01862035
:
260 g/mol
:
115…118 °C/15mmHg
:
>110 °C
:
GHS07
:
Ambient