
2,5-Bis(trifluoromethyl)benzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51012-27-2
:
Formula:C₉H₃F₆N
MDL Number:MFCD03094423
Boiling Pt:194 °C
UN:3276
ADR:6.1,III
:239.12 g/mol
:GHS06
Storage Temperature:Ambient