
2,5-Bis(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:395-64-2
:
Formula:C₉H₄F₆O
MDL Number:MFCD00674087
:242.12 g/mol
Boiling Pt:137 °C
Density:1.468 g/cm³ (25 °C)
Flash Pt:69 °C
:GHS07
Storage Temperature:Refrigerator