
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22767-90-4
:
Formula:C₈H₁₁F₃O₂
MDL Number:MFCD00040841
:196.17 g/mol
Boiling Pt:138…141 °C
Density:1.129 g/cm³ (25 °C)
Flash Pt:41 °C
Storage Temperature:Ambient
EINECS:245-211-0
UN:1224
ADR:3,III
:GHS02