Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Difluoro-2-nitrobenzene
1,3-Difluoro-2-nitrobenzene 98%
1,3-Difluoro-2-nitrobenzene 98%
:
Apollo Scientific
1,3-Difluoro-2-nitrobenzene 98%
:
Apollo Scientific
:
19064-24-5
:
:
C₆H₃F₂NO₂
:
MFCD00192035
:
159.09 g/mol
:
91…92 °C/11mmHg
:
1.503 g/cm³ (25 °C)
:
88 °C
:
242-793-8
:
GHS07
:
Ambient