1,2-Difluoropropane 97%
Catalog # APOSPC7149-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:62126-90-3
Formula:C₃H₆F₂
MDL Number:MFCD00040001