Order Entry
Export
ContactUsLinkComponent
 
Irritant
2,3-Difluoro-4-methoxybenzonitrile 98%
Catalog # APOSPC7137-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Difluoro-4-methoxybenzonitrile 98%
Catalog # APOSPC7137-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :169.13 g/mol
Melting Pt:82…85 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
 :GHS07
CAS Number:256417-12-6
Formula:C₈H₅F₂NO
MDL Number:MFCD03428596