Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-2,6-difluorobenzamide
3-Chloro-2,6-difluorobenzamide 97%
3-Chloro-2,6-difluorobenzamide 97%
3-Chloro-2,6-difluorobenzamide 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Chloro-2,6-difluorobenzamide 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
261762-41-8
:
Formula:
C₇H₄ClF₂NO
MDL Number:
MFCD01631386
:
191.56 g/mol
Melting Pt:
133…136 °C
Storage Temperature:
Ambient